3,5-Bis(trifluoromethyl)benzaldehyde ≥95.0%

Supplier: TCI America

B1751-5G B1751-25G
TCB1751-005GEA 36.34 CAD
TCB1751-005G TCB1751-025G
3,5-Bis(trifluoromethyl)benzaldehyde ≥95.0%
3,5-Bis(trifluoromethyl)benzaldehyde

CAS Number: 401-95-6
MDL Number: MFCD00010206
Molecular Formula: C9H4F6O
Molecular Weight: 242.12
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 171
Flash Point (°C): 70
Specific Gravity (20/20): 1.48

Formula: C₉H₄F₆O
Boiling Pt: 171 °C
Melting Pt: –2 °C
Density: 1.48 (20 °C)
Flash Pt: 70 °C
MDL Number: MFCD00010206
CAS Number: 401-95-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e50572b568b8a9439d8f5eedf9df7b68ec7e5ebf&vid=e50572b568b8a9439d8f5eedf9df7b68ec7e5ebf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR