4-tert-Butylcalix[8]arene ≥98.0% (by HPLC)
Supplier: TCI America
B1750-25G
B1750-5G
TCB1750-025GEA
649.38
CAD
TCB1750-025G
TCB1750-005G
4-tert-Butylcalix[8]arene ≥98.0% (by HPLC)
4-tert-Butylcalix[8]arene
CAS Number: 68971-82-4
MDL Number: MFCD00075392
Molecular Formula: C88H112O8
Molecular Weight: 1297.86
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Product Categories: Materials Chemistry, Macrocycles for Host-Guest Chemistry
Other Product Names / Synonyms: Octa-tert-butyl(octahydroxy)calix[8]arene
Formula:
C₈₈H₁₁₂O₈ MW: 1297.85 g/mol |
MDL Number:
MFCD00075392 CAS Number: 68971-82-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...