1-Butylpyridinium bromide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: N-butylpyridinium bromide, 1-n-Butylpyridinium bromide

B1743-5G B1743-25G
TCB1743-5GEA 53.72 CAD
TCB1743-5G TCB1743-025G
1-Butylpyridinium bromide ≥98.0% (by HPLC, titration analysis)
1-Butylpyridinium bromide

CAS Number: 874-80-6
MDL Number: MFCD00671496
Molecular Formula: C9H14BrN
Molecular Weight: 216.12
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 104

Formula: C₉H₁₄BrN
MW: 216.12 g/mol
Melting Pt: 104 °C
MDL Number: MFCD00671496
CAS Number: 874-80-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c5c156f0e1a622fb6638f6b597b42f786ea7feea&vid=c5c156f0e1a622fb6638f6b597b42f786ea7feea&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR