5-Bromoindole ≥99.0%

Supplier: TCI America

Synonyms: 5-Bromo-1H-indole

B1738-5G B1738-25G
TCB1738-005GEA 45.82 CAD
TCB1738-005G TCB1738-025G
5-Bromoindole ≥99.0%
5-Bromoindole

CAS Number: 10075-50-0
MDL Number: MFCD00005670
Molecular Formula: C8H6BrN
Molecular Weight: 196.05
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 90

Formula: C₈H₆BrN
Melting Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00005670
CAS Number: 10075-50-0
EINECS: 233-208-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=33e24d29fe7a7a517493e21ae2da4845393cf6a2&vid=33e24d29fe7a7a517493e21ae2da4845393cf6a2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR