10-Bromo-1-decanol ≥85.0%
Supplier: TCI America
|
Warning
|
B1730-5G
B1730-25G
TCB1730-005GEA
151.68
CAD
TCB1730-005G
TCB1730-025G
10-Bromo-1-decanol ≥85.0%
10-Bromo-1-decanol
CAS Number: 53463-68-6
MDL Number: MFCD00041681
Molecular Formula: C10H21BrO
Molecular Weight: 237.18
Purity/Analysis Method: >85.0% (GC)
Form: Clear Liquid
Boiling point (°C): 169
Flash Point (°C): 109
Specific Gravity (20/20): 1.16
Storage Temperature: <0°C
Formula:
HO(CH₂)₉CH₂Br Boiling Pt: 169 °C Density: 1.16 (20 °C) Flash Pt: 109 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041681 CAS Number: 53463-68-6 EINECS: 258-572-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...