8-Bromo-1-octanol ≥90.0%

Supplier: TCI America

Synonyms: 8-Bromooctan-1-ol, 1-Bromo-8-hydroxyoctane

B1729-25G B1729-5G
TCB1729-025GEA 636.74 CAD
TCB1729-025G TCB1729-005G
8-Bromo-1-octanol ≥90.0%
8-Bromo-1-octanol

CAS Number: 50816-19-8
MDL Number: MFCD00010388
Molecular Formula: C8H17BrO
Molecular Weight: 209.13
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Flash Point (°C): 110
Specific Gravity (20/20): 1.22
Storage Temperature: 0-10°C

Formula: BrH₂C(CH₂)₆CH₂OH
Density: 1.22 (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00010388
CAS Number: 50816-19-8
EINECS: 256-785-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c2e67cb40176b4901eb7d72761ff653dd7c0f8f&vid=6c2e67cb40176b4901eb7d72761ff653dd7c0f8f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR