4-tert-Butyl-1-bromobenzene ≥96.0%

Supplier: TCI America

Synonyms: 1-Bromo-4-tert-butylbenzene, 2-(4-Bromophenyl)-2-methylpropane, 1-Bromo-4-(tert-butyl)benzene, 4-Bromo-tert-butylbenzene

B1727-25G B1727-100G
TCB1727-025GEA 60.04 CAD
TCB1727-025G TCB1727-100G
4-tert-Butyl-1-bromobenzene ≥96.0%
4-tert-Butyl-1-bromobenzene

CAS Number: 3972-65-4
MDL Number: MFCD00000108
Molecular Formula: C10H13Br
Molecular Weight: 213.12
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 228
Flash Point (°C): 97
Freezing point (°C): 14
Specific Gravity (20/20): 1.25


Other Product Names / Synonyms: 4-tert-Butylbromobenzene

Formula: C₁₀H₁₃Br
MW: 213.12 g/mol
Boiling Pt: 232
Melting Pt: 15.5
Density: 1.25 (20 °C)
Flash Pt: 97 °C
Storage Temperature: Ambient
MDL Number: MFCD00000108
CAS Number: 3972-65-4
EINECS: 223-599-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=29fc420c81fcd7fd3a50ac4ed47d4305ff3c6855&vid=30a0c2844aba419d371b85ab3e0e8334ac9fb6ba&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR