4-Bromo-2-fluorotoluene ≥97.0%

Supplier: TCI America

B1716-5G B1716-25G
TCB1716-005GEA 33.18 CAD
TCB1716-005G TCB1716-025G
4-Bromo-2-fluorotoluene ≥97.0%
4-Bromo-2-fluorotoluene

CAS Number: 51436-99-8
MDL Number: MFCD00013551
Molecular Formula: C7H6BrF
Molecular Weight: 189.03
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 100
Flash Point (°C): 76
Specific Gravity (20/20): 1.51

Formula: C₇H₆BrF
Boiling Pt: 100 °C
Density: 1.51 (20 °C)
Flash Pt: 76 °C
MDL Number: MFCD00013551
CAS Number: 51436-99-8
EINECS: 257-201-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0009b5a9c1ccf5da7f1176705a6b8a3d1fe16def&vid=e28d906fccb287e21f05ef3ec354d48e1251073d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR