(Bromomethyl)cyclohexane ≥97.0%
Supplier: TCI America
|
Danger
|
B1708-500G
B1708-25G
B1708-100G
TCB1708-500GEA
587.76
CAD
TCB1708-500G
TCB1708-25G
76219-350
(Bromomethyl)cyclohexane ≥97.0%
(Bromomethyl)cyclohexane
CAS Number: 2550-36-9
MDL Number: MFCD00001509
Molecular Formula: C7H13Br
Molecular Weight: 177.09
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 94
Flash Point (°C): 70
Specific Gravity (20/20): 1.28
Other Product Names / Synonyms: Cyclohexylmethyl Bromide
Formula:
C₇H₁₃Br Boiling Pt: 94 °C Density: 1.28 (20 °C) Flash Pt: 70 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001509 CAS Number: 2550-36-9 UN: 1993 ADR: 3,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...