1,2-Bis(dimethylsilyl)benzene ≥94.0%
Supplier: TCI America
Synonyms:
1,2-Phenylenebis(dimethylsilane)
CAS Number: 17985-72-7
MDL Number: MFCD00142462
Molecular Formula: C10H18Si2
Molecular Weight: 194.42
Purity/Analysis Method: >94.0% (GC)
Form: Clear Liquid
Boiling point (°C): 128
Flash Point (°C): 70
Specific Gravity (20/20): 0.90
Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis), Silicon Compounds (for Synthesis)
Other Product Names / Synonyms: 1,2-Phenylenebis(dimethylsilane)
Formula:
C₁₀H₁₈Si₂ Boiling Pt: 128 °C Density: 0.90 (20 °C) Flash Pt: 70 °C |
MDL Number:
MFCD00142462 CAS Number: 17985-72-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...