N-Bromophthalimide ≥95.0%

Supplier: TCI America

B1697-5G B1697-25G
TCB1697-5GEA 61.62 CAD
TCB1697-5G TCB1697-025G
N-Bromophthalimide ≥95.0%
N-Bromophthalimide

CAS Number: 2439-85-2
MDL Number: MFCD00005888
Molecular Formula: C8H4BrNO2
Molecular Weight: 226.03
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 207

Formula: C₈H₄BrNO₂
Melting Pt: 207 °C
MDL Number: MFCD00005888
CAS Number: 2439-85-2
EINECS: 219-467-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce5607b5c641239101014334c3c33ef081586d6a&vid=ce5607b5c641239101014334c3c33ef081586d6a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR