4-Bromocinnamic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 3-(4-Bromophenyl)acrylic acid

B1690-25G B1690-5G
TCB1690-025GEA 165.9 CAD
TCB1690-025G TCB1690-5G
4-Bromocinnamic acid ≥98.0% (by GC, titration analysis)
4-Bromocinnamic acid

CAS Number: 1200-07-3
MDL Number: MFCD00004394
Molecular Formula: C9H7BrO2
Molecular Weight: 227.06
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal

Formula: C₉H₇BrO₂
MW: 227.06 g/mol
MDL Number: MFCD00004394
CAS Number: 1200-07-3
EINECS: 214-850-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b7c8e4e34e267c38e8e68d267c1e3de64e1b9872&vid=b7c8e4e34e267c38e8e68d267c1e3de64e1b9872&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR