Bis(triphenylphosphine)palladium(II) chloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

B1667-5G B1667-25G B1667-1G
TCB1667-005GEA 421.54 CAD
TCB1667-005G TCB1667-25G TCB1667-001G
Bis(triphenylphosphine)palladium(II) chloride ≥98.0% (by titrimetric analysis)
Bis(triphenylphosphine)palladium (II) chloride

CAS Number: 13965-03-2
MDL Number: MFCD00009593
Molecular Formula: C36H30Cl2P2Pd
Molecular Weight: 701.90
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Cross-coupling Reaction using Transition Metal Catalysts, Metal Catalysts ([2+2+2] Cycloaddition Reaction)
Other Product Names / Synonyms: Dichlorobis(triphenylphosphine)palladium(II)

Formula: C₃₆H₃₀Cl₂P₂Pd
MDL Number: MFCD00009593
CAS Number: 13965-03-2
EINECS: 237-744-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=efe0e7a11a0d9e162313730478f434d26d5c44af&vid=29292feef786f2633725d108d945badef5adbe14&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR