4-(Benzyloxy)benzoic acid ≥98.0%

Supplier: TCI America

B1665-5G B1665-25G
TCB1665-005GEA 41.08 CAD
TCB1665-005G TCB1665-025G
4-(Benzyloxy)benzoic acid ≥98.0%
4-(Benzyloxy)benzoic acid

CAS Number: 1486-51-7
MDL Number: MFCD00016527
Molecular Formula: C14H12O3
Molecular Weight: 228.25
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 190

Formula: C₁₄H₁₂O₃
MW: 228.25 g/mol
Melting Pt: 190 °C
MDL Number: MFCD00016527
CAS Number: 1486-51-7
EINECS: 216-066-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7853af38e84099bf8247390ce0d0a2491b34be98&vid=7853af38e84099bf8247390ce0d0a2491b34be98&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR