2-Benzyloxyphenol ≥97.0%

Supplier: TCI America

B1661-10G B1661-25G
TCB1661-010GEA 75.84 CAD
TCB1661-010G TCB1661-25G
2-Benzyloxyphenol ≥97.0%
2-Benzyloxyphenol

CAS Number: 6272-38-4
MDL Number: MFCD00002186
Molecular Formula: C13H12O2
Molecular Weight: 200.24
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 215
Flash Point (°C): 113
Specific Gravity (20/20): 1.15


Other Product Names / Synonyms: Catechol Monobenzyl Ether

Formula: C₁₃H₁₂O₂
Boiling Pt: 215 °C
Melting Pt: 38 °C
Density: 1.15 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00002186
CAS Number: 6272-38-4
EINECS: 228-461-5

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR