Fullerene C60 ≥99.0% (by HPLC)

Supplier: TCI America

Synonyms: Fullerene - C60, Buckminster-Fullerene C60, C60 Fullerene, Fullerene-C60, Bucky Ball, Footballen

B1660-100MG B1660-1G
TCB1660-100MGEA 37.92 CAD
TCB1660-100MG TCB1660-001G
Fullerene C60 ≥99.0% (by HPLC)
Fullerene C60

CAS Number: 99685-96-8
MDL Number: MFCD00151408
Molecular Formula: C60
Molecular Weight: 720.66
Purity/Analysis Method: >99.0% (HPLC)
Form: Crystal


Product Categories: Materials Chemistry, Molecular Conductors, Fullerenes (Nanocarbon Materials)
Other Product Names / Synonyms: Buckminsterfullerene C60, C60, Carbon C60

Formula: C₆₀
MW: 720.66 g/mol
Melting Pt: > 280 °C
Density: ∼1.65 g/cm³ (20 °C)
MDL Number: MFCD00151408
CAS Number: 99685-96-8
UN: 1325
ADR: 4.1,III
Merck Index: 13,01447
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f9621e9e818dd79e0860d45aa7fa0184e7954f5d&vid=f9621e9e818dd79e0860d45aa7fa0184e7954f5d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR