Fullerene C60 ≥99.5% (by HPLC), pure
Supplier: TCI America
B1641-1G
B1641-100MG
TCB1641-001GEA
357.08
CAD
TCB1641-001G
TCB1641-100MG
Fullerene C60 ≥99.5% (by HPLC), pure
Fullerene C60
CAS Number: 99685-96-8
MDL Number: MFCD00151408
Molecular Formula: C60
Molecular Weight: 720.66
Purity/Analysis Method: >99.5% (HPLC)
Form: Crystal
Product Categories: Materials Chemistry, Organic Transistor Materials, Molecular Conductors, Solar Cell Research Reagents, Fullerenes (Nanocarbon Materials)
Other Product Names / Synonyms: Buckminsterfullerene C60, C60, Carbon C60
Formula:
C₆₀ Melting Pt: > 280 °C Density: ∼1.65 g/cm³ (20 °C) |
MDL Number:
MFCD00151408 CAS Number: 99685-96-8 UN: 1325 ADR: 4.1,III Merck Index: 13,01447 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...