1-Bromo-4-heptylbenzene ≥96.0% stabilized

Supplier: TCI America

Synonyms: (4-Bromophenyl)heptane

B1608-1G B1608-5G
TCB1608-1GEA 39.5 CAD
TCB1608-1G TCB1608-005G
1-Bromo-4-heptylbenzene ≥96.0% stabilized
1-Bromo-4-heptylbenzene

(stabilized with Copper chip)
CAS Number: 76287-49-5
MDL Number: MFCD00061115
Molecular Formula: C13H19Br
Molecular Weight: 255.20
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 190
Specific Gravity (20/20): 1.15


Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: 4-Heptylbromobenzene

Formula: C₁₃H₁₉Br
MW: 255.2 g/mol
Boiling Pt: 190 °C
Density: 1.15 (20 °C)
MDL Number: MFCD00061115
CAS Number: 76287-49-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dab3043d1e3a6c21ebd6d6c043e44d7adaebd842&vid=dab3043d1e3a6c21ebd6d6c043e44d7adaebd842&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR