1-Bromo-4-pentylbenzene ≥90.0%

Supplier: TCI America

B1605-5G B1605-25G
TCB1605-005GEA 31.6 CAD
TCB1605-005G TCB1605-025G
1-Bromo-4-pentylbenzene ≥90.0%
1-Bromo-4-pentylbenzene

CAS Number: 51554-95-1
MDL Number: MFCD00061113
Molecular Formula: C11H15Br
Molecular Weight: 227.15
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 148
Flash Point (°C): 89
Specific Gravity (20/20): 1.21


Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: 4-Pentylbromobenzene, 1-Amyl-4-bromobenzene

Formula: C₁₁H₁₅Br
Boiling Pt: 148 °C
Density: 1.21 (20 °C)
Flash Pt: 89 °C
MDL Number: MFCD00061113
CAS Number: 51554-95-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d1292c6c125d8efa1c144a3823f1ecab983e05c5&vid=d1292c6c125d8efa1c144a3823f1ecab983e05c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR