4-Benzyloxybenzyl alcohol ≥95.0%

Supplier: TCI America

B1602-25G B1602-5G
TCB1602-25GEA 106.18 CAD
TCB1602-25G TCB1602-5G
4-Benzyloxybenzyl alcohol ≥95.0%
4-Benzyloxybenzyl alcohol

CAS Number: 836-43-1
MDL Number: MFCD00004654
Molecular Formula: C14H14O2
Molecular Weight: 214.26
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 86

Formula: C₁₄H₁₄O₂
Melting Pt: 86 °C
MDL Number: MFCD00004654
CAS Number: 836-43-1
EINECS: 212-649-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b28aa4e3071e41cc5a130fbdd196b98e7952e588&vid=b28aa4e3071e41cc5a130fbdd196b98e7952e588&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR