1-Bromo-4-nonylbenzene ≥95.0%

Supplier: TCI America

Synonyms: (4-Bromophenyl)nonane

B1596-25G B1596-5G
TCB1596-25GEA 1029.84 CAD
TCB1596-25G TCB1596-005G
1-Bromo-4-nonylbenzene ≥95.0%
1-Bromo-4-nonylbenzene

CAS Number: 51554-94-0
MDL Number: MFCD00061116
Molecular Formula: C15H23Br
Molecular Weight: 283.25
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 190
Flash Point (°C): 120
Specific Gravity (20/20): 1.12


Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: 4-Nonylbromobenzene

Formula: C₁₅H₂₃Br
MW: 283.25 g/mol
Boiling Pt: 190 °C
Density: 1.12 (20 °C)
Flash Pt: 120 °C
MDL Number: MFCD00061116
CAS Number: 51554-94-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb7c6d5b6cc1efc748f09f04af8d12bfe9d183a1&vid=eb7c6d5b6cc1efc748f09f04af8d12bfe9d183a1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR