5-Bromo-2'-deoxyuridine (broxuridine) ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

B1575-1G B1575-5G
TCB1575-001GEA 130.19 CAD
TCB1575-001G TCB1575-5G
5-Bromo-2'-deoxyuridine (broxuridine) ≥98.0% (by HPLC, titration analysis)
5-Bromo-2'-deoxyuridine (Broxuridine)

CAS Number: 59-14-3
MDL Number: MFCD00006529
Molecular Formula: C9H11BrN2O5
Molecular Weight: 307.10
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Specific rotation [a]20/D: 22 deg (C=1, H2O)


Product Categories: Bioscience, Pharmacologically-Active Nucleosides and Nucleobases for Research and Experimental Use, Nucleosides and Their Analogs
Other Product Names / Synonyms: 5-Bromo-1-(2-deoxy-beta-D-ribofuranosyl)uracil, 5-BrdU, BUdR

Formula: C₉H₁₁BrN₂O₅
MDL Number: MFCD00006529
CAS Number: 59-14-3
EINECS: 200-415-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f60c43d1aef98eea7763606f5a7990b29b3b746b&vid=f60c43d1aef98eea7763606f5a7990b29b3b746b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR