Bis(triphenylphosphine)nickel(II) chloride ≥96.0%

Supplier: TCI America

Synonyms: Dichlorobis(triphenylphosphine)nickel(II), Bis(triphenylphosphine)nickel(II)chloride

B1571-100G B1571-10G
TCB1571-100GEA 321.55 CAD
TCB1571-100G TCB1571-010G
Bis(triphenylphosphine)nickel(II) chloride ≥96.0%
Bis(triphenylphosphine)nickel (II) chloride

CAS Number: 14264-16-5
MDL Number: MFCD00009592
Molecular Formula: C36H30Cl2NiP2
Molecular Weight: 654.18
Purity/Analysis Method: >96.0% (T)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Activation Reaction of Unactivated Bonds, Cross-coupling Reaction using Transition Metal Catalysts, Metal Catalysts ([2+2+2] Cycloaddition Reaction), Materials Chemistry, Precise Polymerization Reagents
Other Product Names / Synonyms: Dichlorobis(triphenylphosphine)nickel(II)

Formula: C₃₆H₃₀Cl₂NiP₂
MW: 656.19 g/mol
MDL Number: MFCD00009592
CAS Number: 14264-16-5
EINECS: 238-154-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5798ae79f9085fa172f51f825c8d503ce47e02c4&vid=5798ae79f9085fa172f51f825c8d503ce47e02c4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR