2-Bromo-6-methoxynaphthalene ≥98.0%
Supplier: TCI America
B1547-250G
B1547-25G
TCB1547-250GEA
993.82
CAD
TCB1547-250G
TCB1547-025G
2-Bromo-6-methoxynaphthalene ≥98.0%
2-Bromo-6-methoxynaphthalene
CAS Number: 5111-65-9
MDL Number: MFCD00004062
Molecular Formula: C11H9BrO
Molecular Weight: 237.10
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 164
Melting point (°C): 108
Formula:
C₁₁H₉BrO Boiling Pt: 164 °C Melting Pt: 108 °C |
MDL Number:
MFCD00004062 CAS Number: 5111-65-9 EINECS: 225-837-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...