Benzo-18-crown-6 (benzo-1,4,7,10,13,16-hexoxacyclooctadecane) ≥96.0%
Supplier: TCI America
Synonyms:
Benzo-18-crown 6-Ether
B1539-1G
B1539-5G
TCB1539-001GEA
115.34
CAD
TCB1539-001G
TCB1539-005G
Benzo-18-crown-6 (benzo-1,4,7,10,13,16-hexoxacyclooctadecane) ≥96.0%
Benzo-18-crown-6 (Benzo-1,4,7,10,13,16-hexoxacyclooctadecane)
CAS Number: 14098-24-9
MDL Number: MFCD00062741
Molecular Formula: C16H24O6
Molecular Weight: 312.36
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Melting point (°C): 44
Flash Point (°C): 113
Product Categories: Materials Chemistry, Macrocycles for Host-Guest Chemistry
Other Product Names / Synonyms: 2,3-Benzo-1,4,7,10,13,16-hexaoxaoctadec-2-ene
Formula:
C₁₆H₂₄O₆ MW: 312.36 g/mol Melting Pt: 44 °C Flash Pt: 113 °C |
MDL Number:
MFCD00062741 CAS Number: 14098-24-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...