4-Benzylbiphenyl ≥99.0%

Supplier: TCI America

B1519-25G B1519-5G
TCB1519-025GEA 219.78 CAD
TCB1519-025G TCB1519-005G
4-Benzylbiphenyl ≥99.0%
4-Benzylbiphenyl

CAS Number: 613-42-3
MDL Number: MFCD00041831
Molecular Formula: C19H16
Molecular Weight: 244.34
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 286
Melting point (°C): 86


Product Categories: Materials Chemistry, Pressure & Heat Sensitive Recording Materials
Other Product Names / Synonyms: 4-Phenyldiphenylmethane

Formula: C₁₉H₁₆
Boiling Pt: 286 °C
Melting Pt: 86 °C
Density: 1.171 g/cm³ (0 °C)
MDL Number: MFCD00041831
CAS Number: 613-42-3
EINECS: 210-341-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ad0226feb2a0e6fbecab3bfa5f55572ae121f552&vid=ad0226feb2a0e6fbecab3bfa5f55572ae121f552&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR