Ethyl-4-bromobenzoate ≥98.0%
Supplier: TCI America
Synonyms:
4-Bromobenzoic acid ethyl ester, Ethyl 4-bromobenzoate
B1515-25G
B1515-250G
TCB1515-025GEA
83.74
CAD
TCB1515-025G
TCB1515-250G
Ethyl-4-bromobenzoate ≥98.0%
Ethyl 4-bromobenzoate
CAS Number: 5798-75-4
MDL Number: MFCD00016329
Molecular Formula: C9H9BrO2
Molecular Weight: 229.07
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 131
Flash Point (°C): 110
Specific Gravity (20/20): 1.43
Other Product Names / Synonyms: 4-Bromobenzoic Acid Ethyl Ester
Formula:
C₉H₉BrO₂ MW: 229.07 g/mol Boiling Pt: 131 °C Melting Pt: 18 °C Density: 1.43 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00016329 CAS Number: 5798-75-4 EINECS: 227-347-2 Merck Index: 13,01392 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...