1-Bromopyrene ≥95.0%

Supplier: TCI America
Warning

B1495-5G B1495-25G
TCB1495-005GEA 110.6 CAD
TCB1495-005G TCB1495-25G
1-Bromopyrene ≥95.0%
1-Bromopyrene

CAS Number: 1714-29-0
MDL Number: MFCD00015767
Molecular Formula: C16H9Br
Molecular Weight: 281.15
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 255
Melting point (°C): 98

Formula: C₁₆H₉Br
Boiling Pt: 255 °C
Melting Pt: 98 °C
MDL Number: MFCD00015767
CAS Number: 1714-29-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef1246cd447673a30481cc58bf236d15fc304779&vid=ef1246cd447673a30481cc58bf236d15fc304779&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR