Bis(2-hydroxyethyl)amino tris(hydroxymethyl)methane (BIS-TRIS) ≥99.0%

Supplier: TCI America

Synonyms: 2,2-Bis(hydroxymethyl)-2,2',2''-nitrilotriethanol, Bis(2-hydroxyethyl)amino-tris(hydroxymethyl)methane (BIS-TRIS), 2-(Bis(2-hydroxyethyl)amino)-2-(hydroxymethyl)propane-1,3-diol, 2-[Bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol

B1493-250G B1493-25G
TCB1493-250GEA 664.55 CAD
TCB1493-250G TCB1493-025G
Bis(2-hydroxyethyl)amino tris(hydroxymethyl)methane (BIS-TRIS) ≥99.0%
Bis(2-hydroxyethyl)amino-tris(hydroxymethyl)methane (BIS-TRIS)

[Good's buffer component for biological research]
CAS Number: 6976-37-0
MDL Number: MFCD00002853
Molecular Formula: C8H19NO5
Molecular Weight: 209.24
Purity/Analysis Method: >99.0% (HPLC,T)
Form: Crystal


Product Categories: Bioscience, Good's Buffering Agents
Other Product Names / Synonyms: Bis(2-hydroxyethyl)iminotris(hydroxymethyl)methane, Bis-Tris

Formula: C₈H₁₉NO₅
Melting Pt: 102…103 °C
Storage Temperature: Ambient
MDL Number: MFCD00002853
CAS Number: 6976-37-0
EINECS: 230-237-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5710ee8a0f9097cc0827c0ff6d348e06a2c2cad&vid=21a75656b758f3a9099d52aa294486ad5450e1a4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR