4-Bromo-1,3-dimethoxybenzene ≥97.0%
Supplier: TCI America
|
Warning
|
B1481-25G
B1481-5G
TCB1481-025GEA
198.29
CAD
TCB1481-025G
76219-344
4-Bromo-1,3-dimethoxybenzene ≥97.0%
4-Bromo-1,3-dimethoxybenzene
CAS Number: 17715-69-4
MDL Number: MFCD00009844
Molecular Formula: C8H9BrO2
Molecular Weight: 217.06
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 132
Flash Point (°C): 110
Freezing point (°C): 25
Formula:
C₈H₉BrO₂ Boiling Pt: 132 °C Density: 1.507 g/cm³ (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00009844 CAS Number: 17715-69-4 EINECS: 241-717-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...