2-Bromo-3-hydroxypyridine ≥98.0%
Supplier: TCI America
|
Warning
|
Synonyms:
2-Bromo-3-pyridinol
B1478-5G
B1478-25G
TCB1478-005GEA
56.88
CAD
TCB1478-005G
TCB1478-025G
2-Bromo-3-hydroxypyridine ≥98.0%
2-Bromo-3-hydroxypyridine
CAS Number: 6602-32-0
MDL Number: MFCD00006220
Molecular Formula: C5H4BrNO
Molecular Weight: 174.00
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 181
Flash Point (°C): 110
Other Product Names / Synonyms: 2-Bromo-3-pyridinol
Formula:
C₅H₄BrNO MW: 174 g/mol Boiling Pt: 232 Melting Pt: 181 °C Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006220 CAS Number: 6602-32-0 EINECS: 229-547-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...