11-Bromo-1-undecanol ≥97.0%

Supplier: TCI America

B1477-5G B1477-25G
TCB1477-005GEA 33.18 CAD
TCB1477-005G TCB1477-025G
11-Bromo-1-undecanol ≥97.0%
11-Bromo-1-undecanol

CAS Number: 1611-56-9
MDL Number: MFCD00004752
Molecular Formula: C11H23BrO
Molecular Weight: 251.21
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 170
Melting point (°C): 45
Flash Point (°C): 110
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 11-Bromoundecyl Alcohol

Formula: HO(CH₂)₁₀CH₂Br
Boiling Pt: 170 °C
Melting Pt: 45 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004752
CAS Number: 1611-56-9
EINECS: 216-554-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=04b90aa29e16edaedb92af60ec69d50867f4d45b&vid=04b90aa29e16edaedb92af60ec69d50867f4d45b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR