1-Bromo-5-chloropentane ≥98.0%

Supplier: TCI America

B1476-25G B1476-5G
TCB1476-025GEA 79.32 CAD
TCB1476-025G 76219-342
1-Bromo-5-chloropentane ≥98.0%
1-Bromo-5-chloropentane

CAS Number: 54512-75-3
MDL Number: MFCD00001016
Molecular Formula: C5H10BrCl
Molecular Weight: 185.49
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 212
Flash Point (°C): 95
Specific Gravity (20/20): 1.41


Other Product Names / Synonyms: 5-Bromopentyl Chloride, 5-Chloropentyl Bromide

Formula: BrH₂C(CH₂)₃CH₂Cl
MW: 185.49 g/mol
Boiling Pt: 212 °C
Density: 1.41 (20 °C)
Flash Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00001016
CAS Number: 54512-75-3
EINECS: 259-193-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d29f75c0a5352793b8720c5cd7bd50bc563f938a&vid=d29f75c0a5352793b8720c5cd7bd50bc563f938a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR