Bis(phenylthio)methane ≥97.0%

Supplier: TCI America

B1444-5G B1444-25G
TCB1444-005GEA 36.34 CAD
TCB1444-005G TCB1444-025G
Bis(phenylthio)methane ≥97.0%
Bis(phenylthio)methane

CAS Number: 3561-67-9
MDL Number: MFCD00003067
Molecular Formula: C13H12S2
Molecular Weight: 232.36
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 194
Melting point (°C): 37
Flash Point (°C): 110


Product Categories: Synthetic Organic Chemistry, Sulfides, Carbonyl Olefination
Other Product Names / Synonyms: Formaldehyde Diphenyl Mercaptal

Formula: C₁₃H₁₂S₂
Boiling Pt: 194 °C
Melting Pt: 37 °C
Flash Pt: 110 °C
MDL Number: MFCD00003067
CAS Number: 3561-67-9
EINECS: 222-624-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef1ede72f26ca1c284a9791150d6a8856ef9e757&vid=ef1ede72f26ca1c284a9791150d6a8856ef9e757&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR