1-Bromo-2-ethylbenzene ≥98.0%

Supplier: TCI America

B1428-5G B1428-25G
TCB1428-5GEA 77.42 CAD
TCB1428-5G TCB1428-025G
1-Bromo-2-ethylbenzene ≥98.0%
1-Bromo-2-ethylbenzene

CAS Number: 1973-22-4
MDL Number: MFCD00000077
Molecular Formula: C8H9Br
Molecular Weight: 185.06
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 199
Flash Point (°C): 71
Freezing point (°C): -68
Specific Gravity (20/20): 1.36

Formula: C₈H₉Br
MW: 185.06 g/mol
Boiling Pt: 199 °C
Melting Pt: –67.3 °C
Density: 1.36 (20 °C)
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00000077
CAS Number: 1973-22-4
EINECS: 217-823-8
UN: 3082
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c32dc7bdbfbfa2adeb3a0d855095250d8bfb67c4&vid=a532b216836a28af977ff573b52e80c93d42a4ec&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR