2-Bromo-5-chlorotoluene ≥98.0%

Supplier: TCI America
Warning

Synonyms: 1-Bromo-4-chloro-2-methylbenzene

B1427-250G B1427-25G
TCB1427-250GEA 234.79 CAD
TCB1427-250G TCB1427-025G
2-Bromo-5-chlorotoluene ≥98.0%
2-Bromo-5-chlorotoluene

CAS Number: 14495-51-3
MDL Number: MFCD00000602
Molecular Formula: C7H6BrCl
Molecular Weight: 205.48
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 100
Flash Point (°C): 95
Specific Gravity (20/20): 1.56

Formula: C₇H₆BrCl
Boiling Pt: 100 °C
Density: 1.56 (20 °C)
Flash Pt: 95 °C
MDL Number: MFCD00000602
CAS Number: 14495-51-3
EINECS: 238-503-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8012c2bc28cc2acd5bbe9fa046bda161500937fa&vid=46e9423ed7fdd608940ad387e2fa9e4ecdbceb18&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR