4-Bromo-3,5-dihydroxybenzoic acid ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

B1425-250G B1425-25G
TCB1425-250GEA 462.78 CAD
TCB1425-250G TCB1425-025G
4-Bromo-3,5-dihydroxybenzoic acid ≥97.0% (by HPLC, titration analysis)
4-Bromo-3,5-dihydroxybenzoic acid

CAS Number: 16534-12-6
MDL Number: MFCD00002513
Molecular Formula: C7H5BrO4
Molecular Weight: 233.02
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Melting point (°C): 273


Other Product Names / Synonyms: 4-Bromo-alpha-resorcylic Acid

Formula: C₇H₅BrO₄
MW: 233.02 g/mol
Melting Pt: 273 °C
MDL Number: MFCD00002513
CAS Number: 16534-12-6
EINECS: 240-606-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=96c08aeb25cc0b28bb249ab4e6abb17ab7ac36d4&vid=0d9365f669c2bde624480d70c1da827e2988094c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR