3-Bromo-α,α,α,4-tetrafluorotoluene ≥96.0%

Supplier: TCI America
Warning

B1421-5G B1421-25G
TCB1421-5GEA 90.85 CAD
TCB1421-5G TCB1421-001G TCB1421-25G
3-Bromo-α,α,α,4-tetrafluorotoluene ≥96.0%
3-Bromo-α,α,α,4-tetrafluorotoluene

CAS Number: 68322-84-9
MDL Number: MFCD00040938
Molecular Formula: C7H3BrF4
Molecular Weight: 243.00
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 60
Specific Gravity (20/20): 1.72
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Bromo-1-fluoro-4-(trifluoromethyl)benzene, 3-Bromo-alpha,alpha,alpha,4-tetrafluorotoluene

Formula: C₇H₃BrF₄
Boiling Pt: 60 °C
Density: 1.72 (20 °C)
MDL Number: MFCD00040938
CAS Number: 68322-84-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec960373ecb1c727593bba76e16f2ad37d04063d&vid=ec960373ecb1c727593bba76e16f2ad37d04063d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR