4-Bromo-2-chlorobenzoic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
B1407-5G
B1407-1G
TCB1407-5GEA
437.66
CAD
TCB1407-5G
TCB1407-001G
4-Bromo-2-chlorobenzoic acid ≥98.0% (by titrimetric analysis)
4-Bromo-2-chlorobenzoic acid
CAS Number: 59748-90-2
MDL Number: MFCD00040903
Molecular Formula: C7H4BrClO2
Molecular Weight: 235.46
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 173
Formula:
C₇H₄BrClO₂ Melting Pt: 173 °C |
MDL Number:
MFCD00040903 CAS Number: 59748-90-2 EINECS: 261-911-9 UN: 2811 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...