(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl ≥98.0%
Supplier: TCI America
Synonyms:
(R)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, (R)-(+)-BINAP, (R)-BINAP, (R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthalene, (R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl
CAS Number: 76189-55-4
MDL Number: MFCD00010805
Molecular Formula: C44H32P2
Molecular Weight: 622.69
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 242
Specific rotation [a]20/D: 235 deg (C=0.3, Toluene)
Product Categories: Synthetic Organic Chemistry, Asymmetric Synthesis, Cross-coupling Reaction using Transition Metal Catalysts, Ligands, Ligands ([2+2+2]Cycloaddition Reaction)
Other Product Names / Synonyms: (R)-(+)-BINAP
Formula:
C₄₄H₃₂P₂ MW: 622.69 g/mol Melting Pt: 242 °C |
MDL Number:
MFCD00010805 CAS Number: 76189-55-4 Merck Index: 13,01223 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...