(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl ≥98.0%
Supplier: TCI America
Synonyms:
(S)-(-)-BINAP, (S)-BINAP, (S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthalene, (S)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthalene, (S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, (S)-(-)-2,2'-Bis(diphenylphoshino)-1,1'-binaphthyl
CAS Number: 76189-56-5
MDL Number: MFCD00010805
Molecular Formula: C44H32P2
Molecular Weight: 622.69
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 242
Specific rotation [a]20/D: -235 deg (C=0.3, Toluene)
Product Categories: Synthetic Organic Chemistry, Asymmetric Synthesis, Cross-coupling Reaction using Transition Metal Catalysts, Ligands, Ligands ([2+2+2]Cycloaddition Reaction)
Other Product Names / Synonyms: (S)-(-)-BINAP
Formula:
C₄₄H₃₂P₂ MW: 622.69 g/mol Melting Pt: 242 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010805 CAS Number: 76189-56-5 Merck Index: 13,01223 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...