N-(4-Bromobutyl)phthalimide ≥95.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 2-(4-Bromobutyl)isoindoline-1,3-dione

B1394-25G B1394-5G
TCB1394-025GEA 241.71 CAD
TCB1394-025G TCB1394-5G
N-(4-Bromobutyl)phthalimide ≥95.0% (by HPLC, titration analysis)
N-(4-Bromobutyl)phthalimide

CAS Number: 5394-18-3
MDL Number: MFCD00005905
Molecular Formula: C12H12BrNO2
Molecular Weight: 282.14
Purity/Analysis Method: >95.0% (HPLC,T)
Form: Crystal
Melting point (°C): 79

Formula: C₁₂H₁₂BrNO₂
MW: 282.14 g/mol
Melting Pt: 79 °C
MDL Number: MFCD00005905
CAS Number: 5394-18-3
EINECS: 226-401-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=49116b06eac74063a015fa89f77bd3701ba806ed&vid=49116b06eac74063a015fa89f77bd3701ba806ed&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR