4-Bromobenzyl alcohol ≥98.0%

Supplier: TCI America

Synonyms: (4-Bromophenyl)methanol

B1372-25G B1372-100G
TCB1372-025GEA 109.02 CAD
TCB1372-025G TCB1372-100G
4-Bromobenzyl alcohol ≥98.0%
4-Bromobenzyl alcohol

CAS Number: 873-75-6
MDL Number: MFCD00004650
Molecular Formula: C7H7BrO
Molecular Weight: 187.04
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 78

Formula: C₇H₇BrO
MW: 187.04 g/mol
Boiling Pt: 268 °C (1013 hPa)
Melting Pt: 78 °C
MDL Number: MFCD00004650
CAS Number: 873-75-6
EINECS: 212-851-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d9ae217acdeba8cbcfc60fadc3f3c6a24c92ff8f&vid=d9ae217acdeba8cbcfc60fadc3f3c6a24c92ff8f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR