5-Bromo-2-chlorobenzoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
B1344-25G
B1344-10G
TCB1344-025GEA
128.93
CAD
TCB1344-025G
TCB1344-010G
5-Bromo-2-chlorobenzoic acid ≥98.0% (by GC, titration analysis)
5-Bromo-2-chlorobenzoic acid
CAS Number: 21739-92-4
MDL Number: MFCD00002415
Molecular Formula: C7H4BrClO2
Molecular Weight: 235.46
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 158
Formula:
C₇H₄BrClO₂ Boiling Pt: 316 °C (1013 hPa) Melting Pt: 158 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002415 CAS Number: 21739-92-4 EINECS: 244-558-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...