1-Butylpyridinium chloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

B1329-25G B1329-10G
TCB1329-025GEA 297.04 CAD
TCB1329-025G TCB1329-010G
1-Butylpyridinium chloride ≥98.0% (by titrimetric analysis)
1-Butylpyridinium chloride

CAS Number: 1124-64-7
MDL Number: MFCD00060047
Molecular Formula: C9H14ClN
Molecular Weight: 171.67
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 133

Formula: C₉H₁₄ClN
Melting Pt: 133 °C
MDL Number: MFCD00060047
CAS Number: 1124-64-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c2893367c919886d6809d7a5f71f12aceff4f69&vid=2c2893367c919886d6809d7a5f71f12aceff4f69&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR