N-(3-Bromopropyl)phthalimide ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

B1328-25G B1328-500G
TCB1328-025GEA 97.96 CAD
TCB1328-025G TCB1328-500G
N-(3-Bromopropyl)phthalimide ≥98.0% (by HPLC, total nitrogen)
N-(3-Bromopropyl)phthalimide

CAS Number: 5460-29-7
MDL Number: MFCD00005904
Molecular Formula: C11H10BrNO2
Molecular Weight: 268.11
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 73

Formula: C₁₁H₁₀BrNO₂
MW: 268.11 g/mol
Boiling Pt: 433 °C (1013 hPa)
Melting Pt: 73 °C
MDL Number: MFCD00005904
CAS Number: 5460-29-7
EINECS: 226-738-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e38065f30dd45d0a1a9c9c015ee04721b73d2c10&vid=e38065f30dd45d0a1a9c9c015ee04721b73d2c10&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR