α,α,α,α',α',α'-Hexafluoro-3,5-xylidine ≥97.0%

Supplier: TCI America
Warning

Synonyms: 3,5-Di(trifluoromethyl)aniline, 3,5-Bis(trifluoromethyl)aniline

B1324-250G B1324-25G B1324-10G
TCB1324-250GEA 399.74 CAD
TCB1324-250G TCB1324-025G TCB1324-010G
α,α,α,α',α',α'-Hexafluoro-3,5-xylidine ≥97.0%
α,α,α,α',α',α'-Hexafluoro-3,5-xylidine

CAS Number: 328-74-5
MDL Number: MFCD00000394
Molecular Formula: C8H5F6N
Molecular Weight: 229.13
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 85
Flash Point (°C): 98
Specific Gravity (20/20): 1.48


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: alpha,alpha,alpha,alpha',alpha',alpha'-Hexafluoro-3,5-xylidine

Formula: C₈H₅F₆N
MW: 229.12 g/mol
Boiling Pt: 85 °C
Melting Pt: 3 °C
Density: 1.48 (20 °C)
Flash Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD00000394
CAS Number: 328-74-5
EINECS: 206-335-0
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ac9830b1a135e78f46818bc94df41e421ab0471&vid=8ac9830b1a135e78f46818bc94df41e421ab0471&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR