2-Bromo-1-indanol ≥98.0%

Supplier: TCI America

Synonyms: 2-Bromo-1-hydroxyindane

B1300-5G B1300-25G
TCB1300-005GEA 41.08 CAD
TCB1300-005G TCB1300-025G
2-Bromo-1-indanol ≥98.0%
2-Bromo-1-indanol

CAS Number: 5400-80-6
MDL Number: MFCD00003798
Molecular Formula: C9H9BrO
Molecular Weight: 213.07
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 131


Other Product Names / Synonyms: 2-Bromo-1-hydroxyhydrindene, 1-Hydroxy-2-bromoindan

Formula: C₉H₉BrO
Boiling Pt: 290 °C (1013 hPa)
Melting Pt: 131 °C
Storage Temperature: Ambient
MDL Number: MFCD00003798
CAS Number: 5400-80-6
EINECS: 226-442-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b5579477783d2baff9534ca8c2289f156fa54c6e&vid=b5579477783d2baff9534ca8c2289f156fa54c6e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR