(4-Bromophenyl)acetic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B1296-250G B1296-25G
TCB1296-250GEA 221.2 CAD
TCB1296-250G TCB1296-025G
(4-Bromophenyl)acetic acid ≥98.0% (by GC, titration analysis)
(4-Bromophenyl)acetic acid

CAS Number: 1878-68-8
MDL Number: MFCD00004342
Molecular Formula: C8H7BrO2
Molecular Weight: 215.05
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 118

Formula: (C₆H₄Br)CH₂COOH
Boiling Pt: 326 °C (1013 hPa)
Melting Pt: 118 °C
MDL Number: MFCD00004342
CAS Number: 1878-68-8
EINECS: 217-523-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dbf1272a8dae848993b1e3f260f735365fe26f09&vid=dbf1272a8dae848993b1e3f260f735365fe26f09&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR