1-Bromo-2-methylnaphthalene ≥90.0%

Supplier: TCI America
Warning

B1295-5G B1295-25G
TCB1295-005GEA 39.5 CAD
TCB1295-005G TCB1295-025G
1-Bromo-2-methylnaphthalene ≥90.0%
1-Bromo-2-methylnaphthalene

CAS Number: 2586-62-1
MDL Number: MFCD00003871
Molecular Formula: C11H9Br
Molecular Weight: 221.10
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 157
Flash Point (°C): 110
Specific Gravity (20/20): 1.44

Formula: C₁₁H₉Br
MW: 221.1 g/mol
Boiling Pt: 157 °C
Density: 1.44 (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00003871
CAS Number: 2586-62-1
EINECS: 219-966-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=007397406858e22a7ca6d76c285a0462800a6378&vid=007397406858e22a7ca6d76c285a0462800a6378&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR